Learn More
VWR International 315 CREPE FLUTE CIRCLES 100PK
Supplier:  VWR International 28331026
Encompass_Preferred
Filter paper. Grade 315, Crepe Fluted Circles, Folded, Qualitative. pk of 100Particle Retention 25 µmPorosity CoarseFlow Rate Fast
Catalog No. NC1936925