Learn More
Cole-Parmer Masterflex Fitting, 316L Stainless Steel, Straight, Hose Barb to Sanitary Clamp Adapter, 3/8" ID x 3/4" Clamp
Supplier:  Cole-Parmer MFLX3070060
Encompass
These barbed fitting offers tight seal on hoses when used with a hose clamp (not included). Ideal for pharmaceutical industry, and chemical compatibility for use with corrosive acids and bases.
Catalog No. NC0160460
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.