Learn More
Chemglass Life Sciences 32mm O-Ring Flat Flange, Tube ID: 27mm, Tube OD: 32mm, O-Ring Size 222
SDP
Supplier:  Chemglass Life Sciences CG13801
Encompass_Preferred
32mm O-Ring Flat Flange, Tube ID: 27mm, Tube OD: 32mm, O-Ring Size 222. Similar to CG-135 flange but with the mating end of the flange having a precision formed o-ring groove instead of being ground. Joint provides a grease-free vacuum tight seal when used with the CG-139 clamp. O-ring is included with each flange and can be ordered separately, see CG-305.
Catalog No. CG13801