Learn More
Cayman Chemical 32N BIotInylamInoethyldI 10mg
SDP
Supplier:  Cayman Chemical 1646110 MG
Encompass_Preferred
Features a biotin group and propanoic acid separated by a disulfide linkage; allows biotinylation of targets through either a chemical or enzymatic acylation reaction; disulfide bond permits removal of the biotin group with reducing agents
Catalog No. 50-297-1346