Learn More
Cayman Chemical 33-BCRP InhIbItr 5mg
SDP
Supplier:  Cayman Chemical 394035 MG
Encompass_Preferred
A BCRP inhibitor; sensitizes mitoxantrone-resistant H460/MX20 cells to mitoxantrone-induced cell death alone and in combination with UV radiation; sensitizes KB-C2 cells overexpressing P-gp to colchicine-induced cell death; increases intracellular accumulation of mitoxantrone in H460/MX20 cells at 5 µM
Catalog No. 50-291-7887