Learn More
Ace Glass, Inc. Complete 34/45 10mm bearing, straight adapter style with hose connection port
SDP
Supplier:  Ace Glass, Inc. 8066134
Encompass_Preferred
34/45 10mm self-aligning, lubricant-free PTFE Trubore straight adapter style bearing with hose connection port. Use with ACE polished stainless steel or glass 10mm O.D. stir shafts. Can achieve vacuum down to 1mm Hg. Multi-directional.
Catalog No. 50-975-969