Learn More
Chemglass Life Sciences 34/45 Outer Joint, 38mm X 2.0W Tubing
SDP
Supplier:  Chemglass Life Sciences CGB10108
Encompass_Preferred
34/45 Outer Joint, 38mm X 2.0W Tubing. Standard taper outer joint tooled on tubing approximately the same diameter as the top reinforcing ring, affording no hold-up below joint.
Catalog No. 50-153-1893