Learn More
Chemglass Life Sciences 34/45 Outer Joint, Full Length, on 38mm OD x 2mm W tubing
SDP
Supplier:  Chemglass Life Sciences CG10108
Encompass_Preferred
Standard taper outer joint tooled on tubing approximately the same diameter as the top reinforcing ring, affording no hold-up below joint. 34/45 Outer Joint, Full Length, on 38mm OD x 2mm W tubing.
Catalog No. CG10108