Learn More
Ace Glass, Inc. 35/20 10.5mm ID 25cm chromatography column with 200Lreservoir and stopcock.
SDP
Supplier:  Ace Glass, Inc. 590606
Encompass_Preferred
10.5mm I.D., 25cm chromatography column with a 35/20 joint and a 200L integral reservoir. Column has a 2mm bore, 1:5 taper solid PTFE stopcock plug at bottom.
Catalog No. 50-972-577