Learn More
Ace Glass, Inc. Complete 35/25 10mm bearing, straight adapter style
SDP
Supplier:  Ace Glass, Inc. 806660
Encompass_Preferred
35/25 10mm straight-adapter style bearing includes all parts needed. Self-aligning, lubricant free PTFE Trubore bearing is designed for use with 10mm O.D. stainless steel or polished glass shafts.
Catalog No. 50-975-960