Learn More
eMolecules​ 3524-68-3 | Pentaerythritol triacrylate (PETA) | Aladdin Scientific Corporation | MFCD00009607 | 298.291 | C14H18O7 | 96.000 | OCC(COC(=O)C=C)(COC(=O)C=C)COC(=O)C=C | 25g | 845282794
Supplier:  eMolecules​ 845282794
Encompass_Preferred
Pentaerythritol triacrylate (PETA) | Aladdin Scientific Corporation | 3524-68-3 | MFCD00009607 | 298.291 | C14H18O7 | 96.000 | OCC(COC(=O)C=C)(COC(=O)C=C)COC(=O)C=C | 25g | 845282794
Catalog No. NC3967232