Learn More
eMolecules​ 365215-38-9 | 2,6-Bis((R)-4-benzyl-4,5-dihydrooxazol-2-yl)pyridine | Ambeed | MFCD23704922 | 397.478 | C25H23N3O2 | 95.000 | C([C@@H]1COC(=N1)c1cccc(n1)C1=N[C@H](Cc2ccccc2)CO1)c1ccccc1 | 5g | 572827682
Supplier:  eMolecules​ 572827682
Encompass_Preferred
2,6-Bis((R)-4-benzyl-4,5-dihydrooxazol-2-yl)pyridine | Ambeed | 365215-38-9 | MFCD23704922 | 397.478 | C25H23N3O2 | 95.000 | C([C@@H]1COC(=N1)c1cccc(n1)C1=N[C@H](Cc2ccccc2)CO1)c1ccccc1 | 5g | 572827682
Catalog No. 50-004-90679