Learn More
World Precision Instrument NanoFil Needles-NF36BL-2
SDP
Supplier:  World Precision Instrument NF36BL2
Encompass
The replaceable needles used with the NanoFil are available with either blunt or beveled tips. The blunt tip is used for injection into soft tissue and when a uniform solution distribution is needed. The beveled style is used for applications that involve the penetration of a tough tissue.
Catalog No. NC1043078