Learn More
McMaster-Carr 37 Degree Flared Fitting for Steel Tubing
Supplier:  McMaster-Carr 4545K162
Encompass
37 Degree Flared Fitting for Steel Tubing Male Adapter for 1/4" Tube OD x 1/8 BSPT, 4500 PSI
Catalog No. NC1457067