Learn More
Electron Microscopy Sciences Air Sampling 37 mm Cassette Tubing Adapters
SDP
Supplier:  Electron Microscopy Sciences 6523310
Encompass
This Adapter is for connecting the 37 mm cassettes to 1/4" ID tubing.
Catalog No. 50-338-88