Learn More
SKC Inc MCE Filters Preloaded in Cassettes, 0.8 um, 37 mm, 50/pk
SDP
Supplier:  SKC Inc 225301
Encompass
MCE Filter, 0.8 um, 37 mm, preloaded in 3-piece clear plastic cassette, pre-banded, 50/pk. DOES NOT ship to Alaska, Arizona, California, Colorado, Hawaii, Idaho, Nevada, Oregon, Utah, Washington. Wyoming”
Catalog No. NC1890080