Learn More
Tricor Braun 38-400 Continuous Thread Lined Black Phenolic Plastic Closure - P/E Cone Liner - Sold Individually
Supplier:  Tricor Braun 006854
Encompass_Preferred
Offered in a variety of diameters, this black phenolic closure comes with an inserted linear low density polyethylene (LLDPE) cone liner. The cone acts as a plug to help prevent leakage and to protect the product from outside contamination. Most often used for difficult sealing problems in the pharmaceutical and industrial chemical industries.
Catalog No. NC0989514