Learn More
SEQUENCE BIOTECH LLC 384 EZ Tray Adaptor - Turns any Sequence Biotech 96 Channel Benchtop Pipettor into a 384 Channel Pipettor.
Supplier:  SEQUENCE BIOTECH LLC 54563
Encompass_Preferred
SBS Format Accessory developed by Sequence Biotech to turn any Sequence Biotech 96 Channel Benchtop Pipettor into a 384 Channel Pipettor. Simply turn the dial and dispense.
Catalog No. NC3311230