Learn More
Alkali Scientific 384-Well Cell Culture Plate, Clear, Flat Bottom, Non-Treated, Sterile, 1/pk, 100/cs
SDP
Supplier:  Alkali Scientific TL47
Encompass
Total well volume of 112 μL; working well volume of 20 μL – 80 μL. Cell culture microplates are sterile and nonpyrogenic. The 384-well Universal Optics NBS microplate is manufactured using an advanced polymer with high clarity and improved chemical resistant properties.
Catalog No. 50-238-9404