Learn More
Alkali Scientific ASI™ 384 Well Cell Culture Plate, Flat Bottom, 1/pk, 100/cs, White, Tissue Culture Treated
SDP
Supplier:  Alkali Scientific TPW47
Encompass
384 Well Cell Culture Plate Total well volume of 112 μL; working well volume of 20 μL – 80 μL Cell culture microplates are sterile and nonpyrogenic. The 384-well Universal Optics NBS microplate is manufactured using an advanced polymer with high clarity and improved chemical resistant properties. Packing: 
Catalog No. 50-302-8641