Learn More
Analytical Sales & Services Thermo Scientific Nunc 384-Well Storage Plate, DeepWell Plate, Low Binding, Automation Compatible, 225 µL
Supplier:  Analytical Sales & Services 384270
Encompass
The Thermo Scientific Nunc 384-Well Polypropylene DeepWell Storage Plate is designed for reliable storage and processing of samples in scientific applications. Constructed from high-quality virgin polypropylene, it features low binding properties, excellent chemical resistance, and wide temperature stability. Its rounded square well design minimizes wicking and optimizes mixing, ensuring compatibility with automated liquid handling systems.
Catalog No. NC1476174