Learn More
Medchemexpress LLC 3B 20E-24-Norchola-5 1mg | 53495-21-9 | 1 MG

Supplier: Medchemexpress LLC HY1153651MG
(3β,20E)-24-Norchola-5,20(22)-diene-3,23-diol is a steroid-derived allylic alcohol supplied as a white to off-white solid for research use. The compound has formula C23H36O2, molecular weight 344.53, and CAS 53495-21-9. Manufacturer data provide storage and stability guidance for powder and solution forms.
- Steroid-based allylic alcohol used in biochemical research.
- Molecular formula C23H36O2 and molecular weight 344.53.
- CAS number 53495-21-9 for unequivocal identification.
- High purity, ≥98.0%.
- Appearance: white to off-white solid.
- Available in small research quantities such as 1 mg, 5 mg, and 10 mg.
- Storage: powder stable at -20°C for up to 3 years; in solvent stable at -80°C for up to 6 months.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.