Learn More
Medchemexpress LLC 3BrB-PP1 10mg | 956025-99-3 | 10 MG

Supplier: Medchemexpress LLC HY11574110MG
3BrB-PP1 is an ATP-analog research compound used to selectively inhibit analog-sensitive kinases in biochemical and cell-based assays. It is supplied as a 10 mg solid for research use.
- Purity 99.0%
- Molecular formula C16H18BrN5
- Molecular weight 360.25 g/mol
- Soluble in DMSO at 100 mg/mL
- Suitable for in vitro biochemical and cell-based kinase assays
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.