Learn More
Cayman Chemical 3C-B-FLYhydrochlorIde 10mg
SDP
Supplier:  Cayman Chemical 1396310 MG
Encompass_Preferred
An α-methylated analog of 2C-B-FLY, and the dihydrofuran analog of DOB, a potent agonist of 5-HT2A and 5-HT2C; intended for forensic and research purposes
Catalog No. 50-295-4614