Learn More
Aat Bioquest 3-Cyano-7-hydroxycoumarin [CAS 19088-73-4]
SDP
Supplier:  Aat Bioquest 40
Encompass_Preferred
3-Cyano-7-hydroxycoumarin is used as a calibration standard for 3-cyano-7-hydroxycoumarin-based enzyme substrates.
Catalog No. 50-297-9246
 Restricted Item