Learn More
MSC Fas-trak Carpet Brush, Non-Abrasive, 17 in Diameter, for 15-20 in Machines
Supplier:  MSC 35813435
Encompass
This complete universal pad and brush driver set is designed for efficient and effective carpet and floor cleaning. It includes a driver, clutch plate, brush inserts, and four looped, absorbent bonnit pads, offering a powerful solution for removing soil from carpets, profiled tiles, or coated vinyl with low moisture.
  • Universal driver set fits 15 in to 20 in machines.
  • Includes driver, clutch plate, brush inserts, and four looped bonnit pads.
  • Reversible, replaceable, and interchangeable brush inserts.
  • Efficiently cleans up to 3,000 sq ft of commercial carpet per hour.
  • Enables carpets to dry in less than one hour after cleaning.
  • Designed for deep prepping, scrubbing, or stripping floors in half the time.
  • Features non-abrasive white pads for effective cleaning.
  • Utilizes low moisture controllability for deep restorative cleaning.
Catalog No. 50-283-8777