Learn More
ILEX LIFE SCIENCES LLC 3D LIFE 10X CBBUFF PH5.5 600UL
SDP
Supplier:  ILEX LIFE SCIENCES LLC B103
Encompass_Preferred
3-D Life 10x CB-Buffer (pH 5.5), buffer component for fast-gelling 3-D Life Hydrogels (600 ul). INTRO: 3-D Life technology, manufactured in Germany by Cellendes GmbH, is an integrated system for the generation of biomimetic hydrogels used in 3D cell culture. 3-D Life represents the most flexible system for biomimetic hydrogel design. DESCRIPTION: 3-D Life 10x CB-Buffer (pH 5.5) is a 10x concentrated salt and buffer solution which is diluted to physiological strength when used in the 3-D Life system. It is used for the formation of fast gelling hydrogels where it provides a cell environment at pH 5.5, which is compatible with most cell types. In conjunction with 3-D Life 10x CB-Buffer (pH 7.2) it is used to generate pHs of intermediate values to modify the gelation kinetics of fast gelling gels. APPLICATION: Used with other components of the 3-D Life Hydrogel System to establish 3D cell cultures. QUANTITY: 600 µl. Sufficient for at least 6 ml 3-D Life Hydrogel.
Catalog No. 50-273-3956