Learn More
Cayman Chemical 3Ethoy SpINSyn L 17pseud 2.5mg
SDP
Supplier:  Cayman Chemical 279652.5 MG
Encompass_Preferred
An acid degradation product of 3’-ethoxy-5,6-dihydrospinosyn L; has no insecticidal activity against the tobacco budworm up to 64 ppm
Catalog No. 50-293-8876