Learn More
Cygnus Technologies KanCapA™ 3G Protein A ELISA Kit, Highly Sensitive Immunoassay, Single-Step Sample Preparation, 12 kits
Supplier:  Cygnus Technologies F950
Encompass
The KANEKA KanCapA™ 3G Protein A Mix-N-Go™ ELISA Kit offers a precise and efficient method for quantifying recombinant Protein A ligand. It features a unique single-step sample treatment process that simplifies analysis by omitting traditional boiling and centrifugation. With a total assay time of under 2 hours and high sensitivity detecting Protein A levels below 100 pg/mL, this kit enhances performance and reduces complexity, making it suitable for both research and manufacturing applications.
Catalog No. NC2950280