Learn More
Medchemexpress LLC 3H-1,2,4-Triazol-3-one, 5-[[(2R,3S)-2-[(1R)-1-[3,5-bis(trifluoromethyl)phenyl]ethoxy]-3-phenyl-4- | 170729-76-7 | 95.8% | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY130775
Encompass_Preferred
Aprepitant impurity 1, appearing as a white to off-white solid, is an impurity of Aprepitant. This product has specific storage recommendations to maintain its stability and purity.
  • Stable for 3 years when stored as powder at -20°C
  • Stable for 2 years when stored as powder at 4°C
  • Stable for 6 months when stored in solvent at -80°C
  • Stable for 1 month when stored in solvent at -20°C
Catalog No. 50-003-41734