Learn More
Medchemexpress LLC 3H-indolium di-sulfo Cy5 alkyne sodium salt | 1617497-19-4 | 99.0% | 701.83 | C35H40N3NaO7S2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY15170110MG
Encompass_Preferred
DiSulfo-Cy5 alkyne is a di-sulfonated Cy5-family fluorescent dye functionalized with a terminal alkyne for bioorthogonal click chemistry. It is water-soluble and intended for covalent conjugation to azide-bearing biomolecules via copper-catalyzed azide-alkyne cycloaddition (CuAAC), enabling fluorescent labeling for imaging and detection in biochemical assays.
  • Water-soluble, di-sulfonated Cy5 fluorophore.
  • Terminal alkyne for copper-catalyzed azide-alkyne click chemistry.
  • Suitable for labeling proteins, nucleic acids, and small molecules.
  • High purity (~99.0%) with characterized molecular weight.
  • Supplied as a 10 MG vial for laboratory use.
  • Molecular formula C35H40N3NaO7S2 and molecular weight 701.83.
Catalog No. 50-002-86307