Learn More
Medchemexpress LLC 3H-pyrrolo[2,3-d]pyrimidine-4,6-dione derivative (pentafluoropropoxy, trifluoroethoxy phenyl) | 1236767-85-3 | 99.9% | 473.27 g·mol^-1 | C17H11F8N3O4 | 25 MG

Supplier: Medchemexpress LLC HY11742725MG
D5D-IN-326 is a selective, orally active delta-5 desaturase inhibitor provided for research use. It shows low-nanomolar activity against human and rat D5D and is used in biochemical, cellular, and in vivo studies related to fatty acid metabolism and metabolic disease models.
- Selective delta-5 desaturase inhibitor with IC50 ≈ 22 nM (human) and 72 nM (rat).
- High purity (99.9%) suitable for biochemical and cellular assays.
- Supplied as a laboratory-scale powder, typical quantity 25 MG.
- Molecular formula C17H11F8N3O4, molecular weight 473.27 g·mol^-1.
- Recommended storage: powder at -20°C for long-term stability.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.