Learn More
Cayman Chemical 3hydroxy3methylglutarylCo 5mg
SDP
Supplier:  Cayman Chemical 253945 MG
Encompass_Preferred
An intermediate in several metabolic pathways; conversion to mevalonate by HMG-CoA reductase is the rate-limiting first step in the cholesterol biosynthetic pathway; can be cleaved into acetyl-CoA and the ketone body acetoacetate in mitochondria by HMG-CoA lyase; an intermediate in the degradation of leucine
Catalog No. 50-293-5993