Learn More
Cayman Chemical 3Hydroxy4methyl2 5HthIo 10mg
SDP
Supplier:  Cayman Chemical 3629010 MG
Encompass_Preferred
A degradation product of cefaclor; formed under aqueous acidic conditions
Catalog No. 50-291-1746