Learn More
Chemglass Life Sciences Nest 3 L High-Efficiency Erlenmeyer Flask, Polycarbonate, Sterile, Solid Cap (CGN-2165-3LSC)
SDP
Supplier:  Chemglass Life Sciences CGN21653LSC
Encompass
Single-use 3 L high-efficiency Erlenmeyer flask made from medical-grade polycarbonate. Supplied sterile and packaged in nests, the flask is designed for bioprocess and cell culture applications that require clear visibility, reliable sterility, and efficient mixing.
  • Medical-grade polycarbonate construction meeting ISO 10993 and USP <661> requirements.
  • Sterilized by E-beam and non-pyrogenic.
  • Packaged in sterile bags, four per case.
  • Solid cap for secure closure.
  • Baffled bottom to improve agitation and mixing.
  • Clear external graduation scale for easy volume observation.
  • Intended for single-use bioprocess and cell culture workflows.
Catalog No. 50-273-3464