Learn More
Nelson Jameson Company 3M Clean-Trace Water Plus ATP
Supplier:  Nelson Jameson Company 3513039 C
Encompass
Real-time ATP water hygiene test which gives an objective indication of cleanliness in CIP water samples in seconds.
Catalog No. NC1126196