Learn More
Enterprise Technology Solutions 3M EarPlugs,Foam,SlwRecvr,29dB NRR,Cylindr,200Pr/BX,10/CT,YW/BE
SDP
Supplier:  Enterprise Technology Solutions MMM3114101CT
Encompass_Preferred
E-A-R Classic Earplugs set the industry standard for personal safety noise reduction. Soft, slow-recovery foam is moisture-resistant and molds to the shape of your ear canal to provide effective hearing protection. Disposable ear plugs are EARfit testable and feature a Noise Reduction Rating (NRR) of 29 dB. Specialty cylindrical shape is suited to fit most sizes.
Catalog No. 50-008-27951