Learn More
Enterprise Technology Solutions 3M Face Mask,N95 Respirator, Adult, 20/PK, White
SDP
Supplier:  Enterprise Technology Solutions MMM9205P20DC
Encompass_Preferred
Aura N95 Particulate Respirator 9205 provides respiratory protection of at least 95 percent filtration efficiency against certain non-oil-based particles. Lightweight, three-panel construction with soft inner material offers a comfortable, convenient fit. 3M's proprietary filter media, 3M Advanced Electret Media, filters dust and other airborne particles for easy breathing. Curved, low profile conforms well to nose and eye contours, allowing more room for eyewear. Design directs exhaled air away from the nose panel, helping reduce eyewear fogging. Adjustable nose clip helps deliver a custom seal. N95 disposable, particulate respirator comes in individually wrapped package for your convenience and protection from contamination before use.
Catalog No. 50-008-27798