Learn More
SOLVENTUM CORPORATION 3M™ Polisher ST, 4.8 cm × 4.3 cm (1 7/8 × 1 11/16 in.)
Supplier:  SOLVENTUM CORPORATION EMP101STX080R
Encompass
This item must be purchased in multiples of 4 per supplier requirements
3M™ Polisher ST is a fully encapsulated single-use anion exchange (AEX) product intended to reduce host cell protein (HCP) impurities, viruses, and other negatively charged contaminants in flow-through polishing chromatography of biopharmaceutical process streams. 3M Polisher ST is a synthetic, hybrid purifier containing two complementary AEX-functional media: a quaternary ammonium (“Q”) functional nonwoven and a guanidinium (Gu) functional membrane
Catalog No. NC2743264