Learn More
Quality Biological Inc Sodium Acetate (3 M), pH 4.5, 500 mL
SDP
Supplier:  Quality Biological Inc 351309101
Encompass_Preferred
Sodium acetate is a widely used reagent in molecular biology applications. ¿ It is used as a buffer in conjunction with acetic acid, in the buffering range of pH 3.6 ¿ 5.6. ¿ Applications: Sodium acetate is used in the purification and precipitation of nucleic acids, protein crystallization, staining of gels in protein gel electrophoresis, and HPLC. ¿ Storage Conditions: 15¿C to 30¿C ¿ Shipping Condition: ambient temperature ¿ Sterile Filtered ¿ Endotoxin and Mycoplasma Tested ¿ pH: 4.5¿0.1 ¿ Product and/or packaging contains recycled/renewable content ¿ Product and/or packaging is recyclable or reduces waste ¿ Product and/or packaging is safer to human and environmental health ¿ ¿ Max Shelf Life: 36 months
Catalog No. 50-146-839