Learn More
Nelson Jameson Company 3M SPONGE-STICK WITH 10ML
Supplier:  Nelson Jameson Company 3513025 B
Encompass
This item has a minimum order quantity of 2 per supplier requirements. 3M Sponge-Stick with 10 mL Buffered Peptone Water Broth uses simplified packaging to minimize handling and preparation, and reduces waste Benefits: - Stick-mounted sponge design allows the ability to access hard to reach areas without directly handling the sponge. - Stick-mounted sponge design provides the ability to easily snap off the sponge from the handle after swabbing. - Simplified packaging minimizes handling and preparation and reduces waste. - 1.5 x 3 inch biocide-free sponge does not affect organism viability. - 6 x 11 inch sample bag with a 30 oz/0.89 L capacity included for safe, convenient transportation to lab. - 100 per case
Catalog No. NC0865959
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
View Alternate Product