Learn More
AdipoGen 3Me-Sulfobutyl-benzindolium
SDP
Supplier:  AdipoGen CDXT0061GG25
Encompass_Preferred
Chemical. CAS 63149-24-6. Formula C19H23NO3S. MW 345.46. Synthetic. Building block for synthesis. Reagent used for electrophilic phenylation, e.g. of malonates and dithiocarboxylates.
Catalog No. 50-256-0028