Learn More
Chemglass Life Sciences 3mL Clear Graduated High Recovery Vial with 20-400 Black Phenolic Open Top Cap and Gray PTFE/Silicone Septa, 20mm OD x 46mm Height
SDP
Supplier:  Chemglass Life Sciences CG4913V003
Encompass_Preferred
Vials are manufactured from Type 1, Class A Borosilicate glass and come with an open-top cap with PTFE Silicone septa.
Catalog No. 50-009-80625