Learn More
Cayman Chemical 3mthylaPyrroldInopropIop 10mg
SDP
Supplier:  Cayman Chemical 1148510 MG
Encompass_Preferred
A structural isomer of methyl-α-PPP, having the methyl group at the 3 position, rather than the 4 position, of the phenyl group; intended for forensic and research applications
Catalog No. 50-295-1573