Learn More
Cayman Chemical 3Odemethyl ThIocolchIcIn 10mg
SDP
Supplier:  Cayman Chemical 3549710 MG
Encompass_Preferred
A degradation product of thiocolchicoside; formed under acidic and basic conditions
Catalog No. 50-291-0163