Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (3R)-3-Pyrrolidinemethanol | 110013-18-8 | >99.96% | C5H11NO | 500 MG
SDP

Supplier:  Medchemexpress LLC HY60263500MG

Encompass_Preferred

(3R)-3-Pyrrolidinemethanol is a molecular building block that serves as a linker for various chemical syntheses. This compound is typically found as a colorless to off-white liquid.

  • Molecular weight: 101.15
  • Chemical formula: C5H11NO
  • CAS number: 110013-18-8
  • Shipment conditions: Room temperature in continental US
  • Storage conditions: Store at room temperature, protect from light. In solvent: -80°C for 1 year, -20°C for 6 months.
  • Solubility (In Vitro): DMSO: 175 mg/mL (1730.10 mM)

Catalog No. 50-004-34717


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.