Learn More
Medchemexpress LLC (3R)-4-(4-Fluorobenzoyl)-3-methyl-2-piperazinone | 1892599-91-5 | 97.5% | 236.25 | 500 MG
SDP
Supplier:  Medchemexpress LLC HY42895500MG
Encompass_Preferred
(3R)-4-(4-Fluorobenzoyl)-3-methyl-2-piperazinone is a drug intermediate used for the synthesis of various active compounds.
  • Solid state
  • Off-white to light yellow color
  • Store at room temperature for 3 years
  • In solvent, store at -80°C for 2 years or -20°C for 1 year
Catalog No. 50-004-03955