Learn More
Medchemexpress LLC (3R,4R)-1-Benzyl-N,4-dimethylpiperidin-3-amine compound with (3S,4S)-1-benzyl-N,4-dimethyl piperidin-3-amine (1:1) | 95.4% | 436.69 | 25 MG

Supplier: Medchemexpress LLC 25MGHY40349A
Tofacitinib impurity 2 is an impurity of Tofacitinib (HY-40354) and is intended for research use only. This product is not for human use.
- Molecular weight of 436.69
- Formula C28H44N4
- Appearance: liquid
- Purity of 95.42% by LCMS
- Storage at 4°C, protected from light
- In solvent storage at -80°C for 6 months or -20°C for 1 month, protected from light
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.