Learn More
Medchemexpress LLC (3R,4R)-4-(benzyloxy)-3-((tert-butoxycarbonyl)amino)pentanoic acid | 254101-11-6 | MFCD01862935 | 97.0% | 323.38 g/mol | C17H25NO5 | 1 G

Supplier: Medchemexpress LLC HYW1408811G
(3R,4R)-4-(benzyloxy)-3-((tert-butoxycarbonyl)amino)pentanoic acid is a BOC-protected threonine derivative used as a synthetic intermediate in peptide and small-molecule synthesis. It serves as a protected amino acid building block for sequence assembly, derivatization, and medicinal chemistry research.
- BOC-protected threonine derivative for peptide synthesis.
- Useful intermediate for small-molecule and medicinal chemistry applications.
- Molecular formula C17H25NO5 and molecular weight 323.38 g/mol.
- CAS number 254101-11-6 for unambiguous identification.
- Recommended storage: pure form -20°C for long-term, 4°C for short-term.
- Available as a 1 G packaging option.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.