Learn More
Apexbio Technology LLC (-)-[3R,4S]-Chromanol 293B 163163-24-4 10mg
SDP
Supplier:  Apexbio Technology LLC B675310
Encompass_Preferred
(-)-3R4S-Chromanol 293B is a small-molecule inhibitor targeting the cardiac IKs potassium channel It is designed to selectively block the slowly activating delayed rectifier K current (IKs) thereby modulating cardiac repolarization and ventricular action potential duration (-)-3R4S-Chromanol 293B exerts its biological activity primarily through inhibition of IKs channel activity Based on these pharmacological properties (-)-3R4S-Chromanol 293B holds research potential in studies of IKs-channel function cardiac repolarization processes and mechanisms underlying arrhythmias
Catalog No. 50-000-91849